Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RY655 Rat Anti-Mouse IgG1, BD OptiBuild™

Catalog No. BDB772925 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB772925 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB772925 Supplier BD Biosciences Supplier No. 772925
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Mouse Samples. It is validated for Flow Cytometry.

Binds specifically to mouse IgG1 of Igh-Ca and Igh-Cb haplotypes; Does not cross-react with other Ig isotypes; Tandem fluorochrome with excitation maximum at 557 nm and emission maximum at 654 nm; Usable on spectral and conventional cytometers; Designed for excitation by 561 nm yellow-green laser; Contains sodium azide; For research use only
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone X56
Concentration 0.2 mg/mL
Conjugate RY655
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Igh-4; Ighg1; Immunoglobulin heavy constant gamma 1
Host Species Rat
Immunogen Mouse IgG1, κ Antibody
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 16017
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.