Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RY655 Rat Anti-Mouse IgG3, BD OptiBuild™

Catalog No. BDB774465 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB774465 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB774465 Supplier BD Biosciences Supplier No. 774465
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Mouse Samples. It is validated for Flow Cytometry.

The R40-82 antibody specifically recognizes mouse IgG3 of all strains and does not react with other Ig isotypes. The BD Horizon RealYellowTM 655 (RY655) Dye is a tandem fluorochrome with an excitation maximum (Ex Max) at 557 nm and an emission maximum (Em Max) at 654 nm. It can be used on both spectral and conventional cytometers and is designed to be excited by the Yellow-Green laser (561-nm). For conventional instruments with a Yellow-Green laser (561-nm), RY655 can be used as an alternative to PE-Cy5. BD recommends using an optical filter centered near 660 nm (e.g., a 660/30-nm or 670/30-nm bandpass filter) for conventional instruments. For conventional and spectral instruments with both a Blue (488-nm) and Yellow-Green (561-nm) laser and appropriate detectors, it can be used in conjunction with RB670. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to the dye under optimum conditions.
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone R40-82
Concentration 0.2 mg/mL
Conjugate RY655
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Ighg3; Immunoglobulin heavy constant gamma 3
Host Species Rat
Immunogen Mouse Pooled Ig
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.