Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RY703 Mouse Anti-Human IgG, BD Horizon™

Catalog No. BDB571860 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB571860 25 Tests
BDB571809 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB571860 Supplier BD Biosciences Supplier No. 571860
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Human Samples. It is validated for Flow Cytometry.

The G18-145 monoclonal antibody specifically binds to the heavy chain of human immunoglobulin G (IgG) subclasses: IgG1, IgG2, IgG3, and IgG4. It reportedly does not react with heavy chains of other human immunoglobulin isotypes. IgG is a crucial component of the humoral immune response, controlling infection and produced by plasma B-cells. It is found in extracellular fluids like blood, lymph, peritoneal, and cerebrospinal fluids. IgG monomers consist of two light and two heavy chains with two antigen binding sites. Human, mouse, and rat species have four IgG subclasses. The BD Horizon RealYellow 703 (RY703) Dye is a tandem fluorochrome with an excitation maximum (Ex Max) at 557 nm and an emission maximum (Em Max) at 703 nm. It is designed to be excited by the Yellow-Green laser (561 nm) with minimal excitation by the 488 nm Blue laser. For conventional instruments with a Yellow-Green laser (561 nm), RY703 can be used as an alternative to PE-Cy5.5.
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone G18-145
Conjugate RY703
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias IGHG1, IGHG2, IGHG3 and IGHG4
Host Species Mouse
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Target Species Human
Content And Storage The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.