Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RY703 Rat Anti-Mouse I-A/I-E, BD OptiBuild™

Catalog No. BDB771455 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB771455 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB771455 Supplier BD Biosciences Supplier No. 771455
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Secondary Antibody

It is validated for Flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen I-A/I-E
Applications Flow Cytometry
Classification Monoclonal
Clone 2G9
Concentration 0.2 mg/mL
Conjugate RY703
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias H-2I; I-Ad, -Ed; I-Eb and I-Ek MHC class II alloantigens; Ia Ag; MHC II
Host Species Rat
Immunogen BALB/c mouse epidermal Langerhans cells
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 14961, 14969
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.