Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RY743 Mouse Anti-Human IgG2, BD OptiBuild™

Catalog No. BDB772419 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB772419 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB772419 Supplier BD Biosciences Supplier No. 772419
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Human Samples. It is validated for Flow Cytometry.

The HP6002 monoclonal antibody specifically recognizes the IgG2 subclass of human Immunoglobulin G (IgG). IgG2 is the second most abundant IgG subclass in human serum after IgG1, followed by IgG3 and IgG4. It consists of two identical heavy chains (encoded by IGHG2) and two light chains (Igκ or Igλ), linked by disulfide bonds. The HP6002 antibody binds to the CH2 domain of the IgG2 heavy chain and does not cross-react with other immunoglobulin heavy chain subclasses. IgG2 is typically expressed by plasmablasts, plasma cells, memory B cells, and some myeloma or plasmacytoma cells. HP6002 binds to soluble human IgG2, cytophilic IgG2 attached to cells via its Fc region, or human IgG2 antibodies specifically bound to antigens. IgG2 can cross the placenta and is found throughout extravascular fluids. Human IgG2 functions include serving as an antigen receptor for B lymphocytes (transmembrane form) and participating in effector functions such as antibody-dependent neutralization of toxins or microbial infection, and complement fixation (secreted soluble forms).
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone HP6002 (also known as AB4-DH4)
Concentration 0.2 mg/mL
Conjugate RY743
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias IGHG2; Immunoglobulin heavy constant gamma 2
Host Species Mouse
Immunogen Purified Human IgG2 Myeloma Protein
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3501
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.