Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RY743 Mouse Anti-Mouse IgD[a], BD OptiBuild™

Catalog No. BDB772673 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB772673 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB772673 Supplier BD Biosciences Supplier No. 772673
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Mouse Samples. It is validated for Flow Cytometry.

Specifically reacts with the Fc portion of mouse IgD of Igh C[a] haplotype (e.g., AKR, BALB/c, CBA, C3H/He, C58, DBA/1, DBA/2, NZB). Does not react with IgD of Igh-C[b] or Igh-C[e] haplotypes (e.g., C57BL/6, C57BL/10, SJL, A). Effective for detecting cell-surface Ig by immunofluorescent staining with flow cytometric analysis. Reported to stimulate B-cell proliferation. The BD Horizon RealYellowTM 743 (RY743) Dye is a yellow-green tandem fluorochrome optimized for detection on spectral cytometers, designed to be excited by the Yellow-Green laser (561 nm) with minimal excitation by the Blue laser (488 nm). RY743 can be used on a spectral instrument with RY703 and either RY775 or PE-Cy7 for additional color options with Yellow-Green laser excitation. For conventional instruments with a Yellow-Green laser, an optical filter centered near 750 nm (e.g., a 750/60-nm bandpass filter) is recommended.
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone AMS 9.1
Concentration 0.2 mg/mL
Conjugate RY743
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Igh-5a; Igh-5.4; Ighd; Immunoglobulin D
Host Species Mouse
Immunogen BALB/c mouse splenocytes
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 380797
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.