Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RY743 Mouse Anti-Rat IgM, BD OptiBuild™

Catalog No. BDB773290 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB773290 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB773290 Supplier BD Biosciences Supplier No. 773290
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Rat Samples. It is validated for Flow Cytometry.

Reacts with rat IgM monomers and pentamers, but not with other Ig isotypes. Optimized for detection on spectral cytometers with excitation by the Yellow-Green laser (561 nm) and minimal excitation by the Blue laser (488 nm). Excitation maximum at 557 nm and emission maximum at 743 nm.
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone G53-238
Concentration 0.2 mg/mL
Conjugate RY743
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Igh-6; Igh-1a; mu-immunoglobulin; RGD1359202
Host Species Mouse
Immunogen Pooled Rat Ig
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.