Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RY743 Rat Anti-Mouse Ig, λ1, λ2, λ3 Light Chain, BD OptiBuild™

Catalog No. BDB773795 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB773795 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB773795 Supplier BD Biosciences Supplier No. 773795
Add to Cart
Add to Cart

Rat Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Mouse Samples. It is validated for Flow Cytometry.

The R26-46 antibody reacts specifically with mouse Igs bearing λ1, λ2, or λ3 light chains. It does not react with κ light chain or heavy chain. Detection of surface immunoglobulin on Ig λ chain-secreting hybridoma cells has been demonstrated with R26-46 mAb. The BD Horizon RealYellow 743 (RY743) Dye is a tandem fluorochrome with an excitation maximum at 557 nm and an emission maximum at 743 nm. RY743 is optimized for detection on spectral cytometers and designed to be excited by the Yellow-Green laser (561 nm) with minimal excitation by the Blue laser (488 nm).
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone R26-46
Concentration 0.2 mg/mL
Conjugate RY743
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Igl; Ig λ; Immunoglobulin lambda chain complex; Ig λ1/λ2/λ3
Host Species Rat
Immunogen Pooled Mouse Ig
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.