Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RY775 Mouse Anti-Human IgG, BD Horizon™

Catalog No. BDB571720 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB571720 25 Tests
BDB571641 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB571720 Supplier BD Biosciences Supplier No. 571720
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Human Samples. It is validated for Flow Cytometry.

BD Horizon RY775 Mouse Anti-Human IgG is a monoclonal antibody (clone G18-145) that specifically binds to the heavy chain of human immunoglobulin G subclasses: IgG1, IgG2, IgG3, and IgG4. It does not react with heavy chains of other human immunoglobulin isotypes. The antibody is conjugated to the RY775 dye, a tandem fluorochrome with an excitation maximum at 557 nm and an emission maximum at 775 nm. It is designed for use in Flow Cytometry and is routinely tested for reactivity in human samples.
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone G18-145
Conjugate RY775
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias IGHG1, IGHG2, IGHG3 and IGHG4
Host Species Mouse
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Target Species Human
Content And Storage The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.