Learn More
Medchemexpress LLC Ry796 | 1393441-53-6 | 98.6% | 5 MG
SDP
Catalog No. 5000234272
Encompass_Preferred
RY796 is a potent, selective inhibitor of voltage-gated potassium (KV2) channels used in preclinical research; it shows analgesic activity and inhibits KV2.1 and KV2.2 with IC50 values in the submicromolar range.
  • Selective KV2 channel inhibitor (IC50: 0.25 μM for KV2.1, 0.09 μM for KV2.2).
  • Demonstrates analgesic activity in preclinical models.
  • High research-grade purity (98.6%).
  • Soluble in DMSO at 100 mg/mL; in vivo formulations documented.
  • Stable as a powder at standard low-temperature storage conditions.
Catalog No. 50-002-34272 Supplier Medchemexpress LLC Supplier No. HY1200335MG
RY796 is a potent, selective inhibitor of voltage-gated potassium (KV2) channels used in preclinical research; it shows analgesic activity and inhibits KV2.1 and KV2.2 with IC50 values in the submicromolar range.
  • Selective KV2 channel inhibitor (IC50: 0.25 μM for KV2.1, 0.09 μM for KV2.2).
  • Demonstrates analgesic activity in preclinical models.
  • High research-grade purity (98.6%).
  • Soluble in DMSO at 100 mg/mL; in vivo formulations documented.
  • Stable as a powder at standard low-temperature storage conditions.