Learn More
Abcam Ryanodine, Ca2+ release modulator, 1MG
Supplier:  Abcam AB1200831MG
Encompass_Preferred
MW 493.5 Da, Purity >98%. Alkaloid that binds with high affinity to ryanodine receptors to modulate intracellular Ca2+ release. Has complex actions and may stimulate or inhibit Ca2+ release, depending on the concentration used.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0992667