Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ryanodine 99%, Thermo Scientific™

Catalog No. AC328500010
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC328500010 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC328500010 Supplier Thermo Scientific Chemicals Supplier No. 328500010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15662-33-6
Molecular Formula C25H35NO9
Molecular Weight (g/mol) 493.553
InChI Key JJSYXNQGLHBRRK-FXKAWTGQSA-N
Synonym ryanodine
PubChem CID 122173044
SMILES CC1CCC2(C3(CC4(C5(C(C(C3(C5(C2(C1O)O4)O)O)OC(=O)C6=CC=CN6)(C(C)C)O)C)O)C)O

Specifications

Melting Point 215°C to 225°C
Color White
Infrared Spectrum Authentic
Quantity 1 mg
Merck Index 15, 8442
Formula Weight 493.6
Percent Purity ≥98.5%
Physical Form Solid
Chemical Name or Material Ryanodine, 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.