Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ryanodine, MP Biomedicals™

Catalog No. ICN15377001 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15377001 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15377001 Supplier MP Biomedicals, Inc Supplier No. 0215377001

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15662-33-6
Molecular Formula C25H35NO9
Molecular Weight (g/mol) 493.553
MDL Number MFCD00151119
InChI Key JJSYXNQGLHBRRK-FXKAWTGQSA-N
Synonym ryanodine
PubChem CID 122173044
SMILES CC1CCC2(C3(CC4(C5(C(C(C3(C5(C2(C1O)O4)O)O)OC(=O)C6=CC=CN6)(C(C)C)O)C)O)C)O

Specifications

Color White
Quantity 1 mg
Formula Weight 493.5
Physical Form Solid
Chemical Name or Material Ryanodine
Recommended Storage Store at room temperature.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.