Learn More
Aviva Systems Biology Corp RYBP Antibody - middle region (ARP31819_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31819_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RYBP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RYBP. Alternate names: AAP1, DEDAF, YEAF1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54329