Learn More
Aviva Systems Biology Corp RYBP Antibody - C-terminal region (ARP38973_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38973_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RYBP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RYBP. Alternate names: AAP1, DEDAF, YEAF1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56875