Learn More
Instech Rymed InVision Plus clear needle free injection port, 27µl priming volume, sterile, box of 200 pcs
SDP
Catalog No. NC2816874
Encompass
Use these needle-free luer injection ports to create a closed system at the connection point between your infusion tether and the dosing syringe. These ports are a perfect complement to the PinPorts™ in Vascular Access Buttons™ which create a closed system at the animal connection point.
The Rymed InVision-Plus is the only neutral fluid displacement needleless luer connector that combines septum seal integrity and double microbial barrier with a simple, internal cannula design having no moving parts within the fluid pathway.
NOTE: These Rymed connnectors for laboratory animal research only. For human use, purchase from a clinical products supplier.
Features
  • Closed system that permits aseptic technique when connecting a syringe to a luer
  • Straight-through fluid pathway
  • Neutral fluid displacement
  • 100% effective blood clearing
Catalog No. NC2816874 Supplier Instech Supplier No. RYM5001CL
Use these needle-free luer injection ports to create a closed system at the connection point between your infusion tether and the dosing syringe. These ports are a perfect complement to the PinPorts™ in Vascular Access Buttons™ which create a closed system at the animal connection point.
The Rymed InVision-Plus is the only neutral fluid displacement needleless luer connector that combines septum seal integrity and double microbial barrier with a simple, internal cannula design having no moving parts within the fluid pathway.
NOTE: These Rymed connnectors for laboratory animal research only. For human use, purchase from a clinical products supplier.
Features
  • Closed system that permits aseptic technique when connecting a syringe to a luer
  • Straight-through fluid pathway
  • Neutral fluid displacement
  • 100% effective blood clearing