Learn More
Enzo Life Sciences Ryanodine receptor monoclonal antibody (C3-33) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804015C100
Encompass_Preferred
Clone: C3-33. Isotype: IgG1. Immunogen: Dog cardiac RyR (ryanodine receptor). Species reactivity: Human, Mouse, Rat, Rabbit, Guinea pig, Fish, Dog, Chicken, Amphibian. Applications: Flow Cytometry, ICC, IHC (FS), IHC (PS), IP, WB. Formulation: Liquid. Ascites diluted in PBS containing 0.05% sodium azide.
Catalog No. 50-200-9747