Learn More
Perkin Elmer US LLC Ryton Scott Spray Chamber for Avio 200/500
Catalog No. 502103852
Encompass_Preferred
The spray chamber is an integral part of the sample introduction system responsible for filtering the sample mist to permit the appropriate droplet size distribution to reach the plasma. This spray chamber is included in the Scott/Cross-Flow II Sample Introduction Kit (N0790609).
Catalog No. 50-210-3852 Supplier Perkin Elmer US LLC Supplier No. N0790384
The spray chamber is an integral part of the sample introduction system responsible for filtering the sample mist to permit the appropriate droplet size distribution to reach the plasma. This spray chamber is included in the Scott/Cross-Flow II Sample Introduction Kit (N0790609).