Learn More
Abcam Ryuvidine, 1MG
Catalog No. 502672402
Encompass_Preferred
MW 284.3 Da, Purity >98%. Inhibits (IC₅₀ = 500 nM) KMT5A / SETD8 / Pr-SET7 and suppresses H4JK20 monomethylation in vitro. Weakly active against Cdk4 (IC₅₀ = 6 μM). Induces S-phase accumulation in HEK-293 cells. Cytotoxic towards a number of human cancer cell lines. May induce the DNA damage response.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2402 Supplier Abcam Supplier No. AB2542351MG
MW 284.3 Da, Purity >98%. Inhibits (IC₅₀ = 500 nM) KMT5A / SETD8 / Pr-SET7 and suppresses H4JK20 monomethylation in vitro. Weakly active against Cdk4 (IC₅₀ = 6 μM). Induces S-phase accumulation in HEK-293 cells. Cytotoxic towards a number of human cancer cell lines. May induce the DNA damage response.
The product is subject to the following: Abcam Restricted Use Statement