Learn More
Medchemexpress LLC (S)-1,1'-bi-2,2'-naphthol | 18531-99-2 | MFCD00004068 | 100.0% | 286.32 | C20H14O2 | 1 ML
SDP
Catalog No. 5000301339
Encompass_Preferred
(S)-1,1'-Bi-2,2'-naphthol is a chiral diol used as a ligand and chiral auxiliary in asymmetric synthesis and catalysis. Supplied as a 10 mM solution in DMSO for research use, it facilitates formation of chiral catalysts, supports enantioselective transformations, and aids method development in organic synthesis.
  • Chiral diol useful as a ligand for asymmetric catalysis.
  • High chemical purity suitable for research and analytical applications.
  • Provided as a 10 mM solution ready for reconstitution or direct use.
  • Characterized by supporting analytical data such as COA, 1H NMR, and RP-HPLC.
Catalog No. 50-003-01339 Supplier Medchemexpress LLC Supplier No. HYW017765A10MM/1ML
(S)-1,1'-Bi-2,2'-naphthol is a chiral diol used as a ligand and chiral auxiliary in asymmetric synthesis and catalysis. Supplied as a 10 mM solution in DMSO for research use, it facilitates formation of chiral catalysts, supports enantioselective transformations, and aids method development in organic synthesis.
  • Chiral diol useful as a ligand for asymmetric catalysis.
  • High chemical purity suitable for research and analytical applications.
  • Provided as a 10 mM solution ready for reconstitution or direct use.
  • Characterized by supporting analytical data such as COA, 1H NMR, and RP-HPLC.