Learn More
Medchemexpress LLC (S)-1,1'-bi-2,2'-naphthol | 18531-99-2 | MFCD00004068 | 100.0% | 286.32 | C20H14O2 | 25 G
SDP
Catalog No. 5000302712
Encompass_Preferred
(S)-1,1'-Bi-2,2'-naphthol (S)-BINOL is a chiral diol used widely as a ligand and chiral auxiliary in asymmetric synthesis and catalysis. It is supplied as a white to off-white solid with high purity, intended for research use in organic synthesis, catalyst development, and related life-science applications.
  • Chiral ligand for asymmetric catalysis and ligand design.
  • High purity suitable for research and synthesis.
  • Solid, white to off-white appearance for easy handling.
  • Stable under recommended storage conditions for prolonged shelf life.
  • Useful as a starting material or chiral auxiliary in academic and industrial research.
Catalog No. 50-003-02712 Supplier Medchemexpress LLC Supplier No. 25GHYW017765A
(S)-1,1'-Bi-2,2'-naphthol (S)-BINOL is a chiral diol used widely as a ligand and chiral auxiliary in asymmetric synthesis and catalysis. It is supplied as a white to off-white solid with high purity, intended for research use in organic synthesis, catalyst development, and related life-science applications.
  • Chiral ligand for asymmetric catalysis and ligand design.
  • High purity suitable for research and synthesis.
  • Solid, white to off-white appearance for easy handling.
  • Stable under recommended storage conditions for prolonged shelf life.
  • Useful as a starting material or chiral auxiliary in academic and industrial research.