Learn More
Cayman Chemical S- 1 1-BI-2-naphthol 1g
SDP
Catalog No. 502896844
Encompass_Preferred
A chiral auxiliary for asymmetric ketone reduction, in particular 15-keto PG intermediates to 15(S)-alcohols
Catalog No. 50-289-6844 Supplier Cayman Chemical Supplier No. 700521 G
A chiral auxiliary for asymmetric ketone reduction, in particular 15-keto PG intermediates to 15(S)-alcohols