Learn More
Cayman Chemical S- 1 1-BI-2-naphthol 25g
SDP
Catalog No. 502893679
Encompass_Preferred
A chiral auxiliary for asymmetric ketone reduction, in particular 15-keto PG intermediates to 15(S)-alcohols
Catalog No. 50-289-3679 Supplier Cayman Chemical Supplier No. 7005225 G
A chiral auxiliary for asymmetric ketone reduction, in particular 15-keto PG intermediates to 15(S)-alcohols