Learn More
Strem, An Ascensus Company CAS# 18531-99-2. 5g. S-(-)-1,1'-Bi-2-naphthol, 99% (S)-BINOL. MFCD00004068
SDP
Catalog No. 5090109211
Encompass
CAS# 18531-99-2. 5g. S-(-)-1,1'-Bi-2-naphthol, 99% (S)-BINOL. MFCD00004068. Molecular Weight: 286.33. Molecular Formula: HOC10H6-C10H6OH. Color/form: white to off-white pwdr. Strem# 08-1001. http://www.strem.com/catalog/v/08-1001/
Catalog No. 50-901-09211 Supplier Strem, An Ascensus Company Supplier No. 0810015G
Add to Cart
Add to Cart
CAS# 18531-99-2. 5g. S-(-)-1,1'-Bi-2-naphthol, 99% (S)-BINOL. MFCD00004068. Molecular Weight: 286.33. Molecular Formula: HOC10H6-C10H6OH. Color/form: white to off-white pwdr. Strem# 08-1001. http://www.strem.com/catalog/v/08-1001/