Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(-)-1-(1-Naphthyl)Ethylamine

Catalog No. 2374505G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-745-05G 5 g
23-745-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 23-745-05G Supplier Sigma Aldrich Supplier No. 2374505G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Employed in a preparation of a chiral thiourea solvating agent useful for enantiodiscrimination of +a-hydroxy and +a-amino acids by 1H NMR.

Specifications

CAS 10420-89-0
Density 1.067 g/mL at 20°C
Boiling Point 153°C (11 mmHg, lit.)
Molecular Formula C12H13N
Refractive Index n20/D 1.623
Linear Formula C10H7CH(CH3)NH2
MDL Number MFCD00064179
Quantity 5 g
Synonym (S)-(-)-alpha-Methyl-1-naphthalenemethylamine
Molecular Weight (g/mol) 171.24 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.