Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-1-(1-Naphthyl)ethylamine, ChiPros 99+%, ee 99+%, Thermo Scientific™

Catalog No. AAL1930106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19301-06 5 g
AAL19301-14 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL19301-06 Supplier Thermo Scientific Chemicals Supplier No. L1930106

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10420-89-0
Molecular Formula C12H14N
Molecular Weight (g/mol) 172.25
MDL Number MFCD00064179
InChI Key RTCUCQWIICFPOD-VIFPVBQESA-O
Synonym s---1-1-naphthyl ethylamine, s-1-naphthalen-1-yl ethanamine, 1s-1-naphthalen-1-yl ethan-1-amine, s---alpha-1-naphthyl ethylamine, s-1-1-naphthyl ethylamine, 1s-1-1-naphthyl ethanamine, s---1-naphthyl ethylamine, s-alpha-methyl-1-naphthalenemethanamine, 1s-1-naphthalen-1-yl ethanamine
PubChem CID 66325
SMILES C[C@H]([NH3+])C1=C2C=CC=CC2=CC=C1

Specifications

Density 1.06
Boiling Point 152°C to 153°C (11 mmHg)
Flash Point >110°C (230°F)
Odor Amine-like
Refractive Index 1.623
Quantity 5 g
UN Number UN2922
Beilstein 2208024
Sensitivity Air Sensitive
Optical Rotation −55° (c=2 in Ethanol)
IUPAC Name (1S)-1-(naphthalen-1-yl)ethan-1-aminium
Formula Weight 171.24
Percent Purity ≥99%
Assay ee ≥99%
Chemical Name or Material (S)-(-)-1-(1-Naphthyl)ethylamine
Show More Show Less
Hazard Category H301-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318-H312
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 600-536-0
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.