Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(+)-1,2,3,4-Tetrahydro-1-Naphthylamine

Catalog No. 6687965G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-879-65G 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 66-879-65G Supplier Sigma Aldrich Supplier No. 6687965G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine is a chiral amine that can be used:To prepare a 2-pyridylmethylene-(S)-(+)-1,2,3,4-tetrahydro-1-naphthylamine containing pentanuclear trigonal bipyramidal [Co3Fe2] complex, which exhibits electron-transfer-coupled spin transition (ETCST) properties.To prepare imine-substituted thiophene Schiff base compounds.To derivatize macrocyclic diarylheptanoid compounds of biological importance.Tetralin is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 7789-29-9
Density 1.136 g/mL at 25°C
Molecular Formula HF2K
Refractive Index n20/D 1.565
Linear Formula KHF2
MDL Number MFCD00011403
Quantity 100 mL
Synonym (S)-(+)-1-AminoTetralin(TM); (S)-(+)-1-Aminotetraline
Molecular Weight (g/mol) 78.1 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.