Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(-)-1,2,4-Butanetriol

Catalog No. 2966785G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-667-85G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 29-667-85G Supplier Sigma Aldrich Supplier No. 2966785G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(-)-1,2,4-Butanetriol can be prepared via reduction of (S)-malic acid in the presence of borane-dimethyl sulfide.(S)-(-)-1,2,4-Butanetriol may be used as a starting material in the enantioselective total syntheses of (+)-azimine and (+)-carpaine. It can also be used to prepare the following organic building blocks:(+)-3,4-epoxy-1-butanol(2S,4S)-4-(hydroxymethyl)-2-ferrocenyl-1,3-dioxane(S)-1,2,4-triacetoxybutane via acetylation with acetic anhydride(S)-1,2,4-tris-(3,5-dinitrobenzoy1oxy)butane via esterification with 3,5-dinitrobenzoyl chloride

Specifications

CAS 42890-76-6
Density 1.19 g/mL at 25°C
Boiling Point 150°C (0.04 mmHg, lit.)
Molecular Formula C4H10O3
Linear Formula HOCH2CH2CH(OH)CH2OH
Refractive Index n20/D 1.475
MDL Number MFCD00063213
Quantity 5 g
Synonym (2S)-(-)-Butane-1,2,4-triol; (S)-(-)-1,2,4-Trihydroxybutane
Molecular Weight (g/mol) 106.12 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.