Learn More
Medchemexpress LLC (S)-1-((2',6-bis(difluoromethyl)-[2,4'-bipyridin]-5-yl)oxy)-2,4-dimethylpentan-2-amine | 1815613-42-3 | 98.8% | C19H23F4N3O | 10MG
SDP
Catalog No. 5000261307
Encompass_Preferred
A small-molecule, CNS-penetrant AAK1 inhibitor supplied as a solid for research use. It is characterized for preclinical in vitro and in vivo studies, with documented purity, solubility in DMSO, and recommended storage conditions for solid and solution forms.
  • CNS-penetrant AAK1 inhibitor suitable for neuropathic pain research.
  • High purity (≈98.8%) appropriate for analytical and pharmacology studies.
  • Off-white to light yellow solid with defined storage recommendations.
  • Soluble in DMSO at high concentration; example in vivo formulations provided.
  • Supplied for research use only; not for human therapeutic use.
Catalog No. 50-002-61307 Supplier Medchemexpress LLC Supplier No. HY13482910MG
A small-molecule, CNS-penetrant AAK1 inhibitor supplied as a solid for research use. It is characterized for preclinical in vitro and in vivo studies, with documented purity, solubility in DMSO, and recommended storage conditions for solid and solution forms.
  • CNS-penetrant AAK1 inhibitor suitable for neuropathic pain research.
  • High purity (≈98.8%) appropriate for analytical and pharmacology studies.
  • Off-white to light yellow solid with defined storage recommendations.
  • Soluble in DMSO at high concentration; example in vivo formulations provided.
  • Supplied for research use only; not for human therapeutic use.