Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(-)-1,2-Epoxybutane

Catalog No. 5323635G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-236-35G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 53-236-35G Supplier Sigma Aldrich Supplier No. 5323635G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(-)-1,2-Epoxybutane can be used:As a starting material to prepare (+)- and (-)-homononactic acids, which are used as intermediates in the total synthesis of a cyclic antibiotic tetranactin.To prepare a chiral phosphorus synthon, which is applicable in the synthesis of phytoprostane B1 type I.To prepare Eu3+-based precatalysts applicable in the Mukaiyama Aldol reaction in water.Manufactured under license by Sterling Pharma Solutions Limited, using Jacobsen HKR technology.

Specifications

CAS 86-58-8
Melting Point >300°C
Molecular Formula C9H8BNO2
Linear Formula C9H8BNO2
Refractive Index n20/D 1.386
MDL Number MFCD01114698
Quantity 1 g
Synonym 8-Quinolineboronic acid
Molecular Weight (g/mol) 172.98 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.