Learn More
Medchemexpress LLC (S)-(+)-1,2-Propanediol | 4254-15-3 | 99.7% | C3H8O2 | 1 KG
SDP
Catalog No. 5000433093
Encompass_Preferred
(S)-(+)-1,2-Propanediol is an endogenous metabolite. It can be utilized as an organic solvent and diluent in drug preparations, as well as a chiral synthetic intermediate and a substrate for enzyme research.
  • Endogenous metabolite
  • Organic solvent
  • Diluent in drug preparations
  • Chiral synthetic intermediate
  • Substrate for enzyme research
Catalog No. 50-004-33093 Supplier Medchemexpress LLC Supplier No. HY793341KG
(S)-(+)-1,2-Propanediol is an endogenous metabolite. It can be utilized as an organic solvent and diluent in drug preparations, as well as a chiral synthetic intermediate and a substrate for enzyme research.
  • Endogenous metabolite
  • Organic solvent
  • Diluent in drug preparations
  • Chiral synthetic intermediate
  • Substrate for enzyme research