Learn More
Medchemexpress LLC (S)-(+)-1,2-propanediol | 4254-15-3 | 99.97% | C3H8O2 | 10 MM 1 ML
SDP
Catalog No. 5000432362
Encompass_Preferred
(S)-(+)-1,2-Propanediol is an endogenous metabolite. It can be used as an organic solvent and diluent in drug preparations. It can also be used as a chiral synthetic intermediate and a substrate for enzyme research. It is for research use only.
  • Endogenous metabolite
  • Organic solvent and diluent in drug preparations
  • Chiral synthetic intermediate
  • Substrate for enzyme research
  • For research use only
Catalog No. 50-004-32362 Supplier Medchemexpress LLC Supplier No. HY7933410MM/1ML
(S)-(+)-1,2-Propanediol is an endogenous metabolite. It can be used as an organic solvent and diluent in drug preparations. It can also be used as a chiral synthetic intermediate and a substrate for enzyme research. It is for research use only.
  • Endogenous metabolite
  • Organic solvent and diluent in drug preparations
  • Chiral synthetic intermediate
  • Substrate for enzyme research
  • For research use only