Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-1,2-Propanediol, 99%, Thermo Scientific™

Catalog No. AC218100010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC218100010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC218100010 Supplier Thermo Scientific Chemicals Supplier No. 218100010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4254-15-3
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.10
MDL Number MFCD00004539
InChI Key DNIAPMSPPWPWGF-UHFFFAOYNA-N
Synonym s-+-1,2-propanediol, s-propane-1,2-diol, s-1,2-propanediol, 2s-propane-1,2-diol, s-propylene glycol, s-+-propylene glycol, s-+-1,2-dihydroxypropane, propylene glycol #, pgo
PubChem CID 439846
ChEBI CHEBI:29002
IUPAC Name propane-1,2-diol
SMILES CC(O)CO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3CH(OH)CH2OH
Quantity 1 g
Merck Index 15, 7968
Specific Rotation Condition +16.80° (20°C c=neat)
Formula Weight 76.09
Chemical Name or Material (S)-(+)-1, 2-Propanediol, 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.