Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(+)-1-(2-Pyrrolidinylmethyl)Pyrrolidine

Catalog No. 324450250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-445-0250MG 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 32-445-0250MG Supplier Sigma Aldrich Supplier No. 324450250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(+)-1-(2-Pyrrolidinylmethyl)pyrrolidine can be used as an organocatalyst in the asymmetric synthesis of different optically active organic building blocks via Aldol condensation and Mannich reactions . It is also used as a catalyst to prepare enantioselective Michael adducts by reacting alkylidene malonates with unactivated ketones.

Specifications

CAS 23364-44-5
Melting Point 142°C to 144°C
Molecular Formula C14H15NO
Linear Formula C6H5CH(NH2)CH(C6H5)OH
Refractive Index n20/D 1.4871
MDL Number MFCD00074960
Quantity 5 g
Molecular Weight (g/mol) 213.28 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.