Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-1-(2-Pyrrolidinylmethyl)pyrrolidine 98.0+%, TCI America™
SDP

Catalog No. P12411G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12411G 1 g
P12415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12411G Supplier TCI America Supplier No. P12411G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51207-66-0
Molecular Formula C9H18N2
Molecular Weight (g/mol) 154.257
MDL Number MFCD00013441
InChI Key YLBWRMSQRFEIEB-VIFPVBQESA-N
PubChem CID 853152
IUPAC Name 1-[[(2S)-pyrrolidin-2-yl]methyl]pyrrolidine
SMILES C1CCN(C1)CC2CCCN2

Specifications

Color Yellow
Boiling Point 84°C
Quantity 1 g
Formula Weight 154.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (S)-(+)-1-(2-Pyrrolidinylmethyl)pyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.