Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol 98.0+%, TCI America™
SDP

Catalog No. B52441G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52441G 1 g
B52445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B52441G Supplier TCI America Supplier No. B52441G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 225920-05-8
Molecular Formula C10H8F6O
Molecular Weight (g/mol) 258.163
MDL Number MFCD03093010
InChI Key MMSCIQKQJVBPIR-YFKPBYRVSA-N
Synonym (S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethyl Alcohol, (S)-3,5-Bis(trifluoromethyl)-alpha-methylbenzyl Alcohol
PubChem CID 9816575
IUPAC Name (1S)-1-[3,5-bis(trifluoromethyl)phenyl]ethanol
SMILES CC(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)O

Specifications

Melting Point 54°C
Color White
Quantity 1 g
Formula Weight 258.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.