Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-1-(3-Methoxyphenyl)ethyl isocyanate, 97%, Thermo Scientific™

Catalog No. AAL2033103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2033103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL2033103 Supplier Thermo Scientific Chemicals Supplier No. L2033103

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 745784-08-1
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.203
MDL Number MFCD05664128
InChI Key YNWCQMWZIOMQOP-QMMMGPOBSA-N
Synonym s---1-3-methoxyphenyl ethyl isocyanate, 1-1s-1-isocyanatoethyl-3-methoxybenzene, 1-1s-1-isocyanatoethyl-3-methoxy-benzene, s---1-3-methoxyphenyl ethylisocyanate, benzene,1-1s-1-isocyanatoethyl-3-methoxy-9ci, benzene, 1-1s-1-isocyanatoethyl-3-methoxy-9ci
PubChem CID 7018285
IUPAC Name 1-[(1S)-1-isocyanatoethyl]-3-methoxybenzene
SMILES CC(C1=CC(=CC=C1)OC)N=C=O

Specifications

Quantity 1 g
UN Number UN2206
Optical Rotation −18° (c=3 in Chloroform)
Formula Weight 177.2
Percent Purity 97%
Chemical Name or Material (S)-(-)-1-(3-Methoxyphenyl)ethyl isocyanate
Hazard Category Danger
Hazard Statement GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Recommended Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.