Learn More
Medchemexpress LLC (S)-1-(4-bromo-6-pyrimidinyl)-N-(2,4-dichlorobenzyl)-2-(pyrrolidin-1-yl)ethan-1-amine hydrochloride | 885104-09-6 | 99.9% | 438.58 g·mol⁻¹ | C15H16BrCl3N4 | 50 MG
SDP
Catalog No. 5000240362
Encompass_Preferred
LY2389575 hydrochloride is the hydrochloride salt of a small-molecule negative allosteric modulator of the metabotropic glutamate receptor 3 (mGlu3), intended for preclinical research. It is supplied as a solid powder with high purity and defined storage conditions for long-term stability.
  • Negative allosteric modulator of mGlu3 for preclinical research
  • High purity (99.9%) suitable for analytical and biological assays
  • Solid powder form for convenient handling and formulation
  • Defined storage stability: powder -20°C up to 3 years or 4°C up to 2 years
  • Molecular weight 438.58 g·mol⁻¹ and formula C15H16BrCl3N4
Catalog No. 50-002-40362 Supplier Medchemexpress LLC Supplier No. HY10750950MG
LY2389575 hydrochloride is the hydrochloride salt of a small-molecule negative allosteric modulator of the metabotropic glutamate receptor 3 (mGlu3), intended for preclinical research. It is supplied as a solid powder with high purity and defined storage conditions for long-term stability.
  • Negative allosteric modulator of mGlu3 for preclinical research
  • High purity (99.9%) suitable for analytical and biological assays
  • Solid powder form for convenient handling and formulation
  • Defined storage stability: powder -20°C up to 3 years or 4°C up to 2 years
  • Molecular weight 438.58 g·mol⁻¹ and formula C15H16BrCl3N4