Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-1-(4-Fluorophenyl)ethyl isocyanate, 95%

Catalog No. AAL2023103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2023103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL2023103 Supplier Thermo Scientific Chemicals Supplier No. L2023103
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 745783-74-8 | C9H8FNO | 165.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 745783-74-8
Molecular Formula C9H8FNO
Molecular Weight (g/mol) 165.167
MDL Number MFCD05664076
InChI Key MUHCZVYPBWOTTJ-ZETCQYMHSA-N
Synonym s---1-4-fluorophenyl ethyl isocyanate, 1-fluoro-4-1s-1-isocyanatoethyl benzene, benzene, 1-fluoro-4-1s-1-isocyanatoethyl-9ci
PubChem CID 7018264
IUPAC Name 1-fluoro-4-[(1S)-1-isocyanatoethyl]benzene
SMILES CC(C1=CC=C(C=C1)F)N=C=O

Specifications

Refractive Index 1.498
Quantity 1 g
UN Number UN2206
Sensitivity Moisture sensitive
Formula Weight 165.17
Percent Purity 95%
Chemical Name or Material (S)-(-)-1-(4-Fluorophenyl)ethyl isocyanate
Hazard Category H302+H332-H315-H317-H319-H335
Hazard Statement GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.