Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-1-(6-(4-fluoro-1H-pyrazol-1-yl)pyridin-3-yl)ethanamine dihydrochloride | 1980023-97-9 | 98.3% | 279.15 | C10H13Cl2FN4 | 100 MG
SDP

Catalog No. 5000210209
Encompass_Preferred

(S)-1-(6-(4-fluoro-1H-pyrazol-1-yl)pyridin-3-yl)ethanamine dihydrochloride is a chiral drug intermediate used in the synthesis of active pharmaceutical compounds.

  • Chiral dihydrochloride salt suitable for synthetic applications.
  • High purity (98.3%) for research use.
  • Molecular formula C10H13Cl2FN4 and molecular weight 279.15.
  • Available in milligram- to gram-scale pack sizes for development and scale-up.

Catalog No. 50-002-10209 Supplier Medchemexpress LLC Supplier No. HYW068075A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-1-(6-(4-fluoro-1H-pyrazol-1-yl)pyridin-3-yl)ethanamine dihydrochloride is a chiral drug intermediate used in the synthesis of active pharmaceutical compounds.

  • Chiral dihydrochloride salt suitable for synthetic applications.
  • High purity (98.3%) for research use.
  • Molecular formula C10H13Cl2FN4 and molecular weight 279.15.
  • Available in milligram- to gram-scale pack sizes for development and scale-up.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.