Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-1-(6-(4-fluoro-1H-pyrazol-1-yl)pyridin-3-yl)ethanamine dihydrochloride | 1980023-97-9 | MFCD32670786 | 98.3% | 279.15 | C10H13Cl2FN4 | 1 G
SDP

Catalog No. 5000210633
Encompass_Preferred

A chiral dihydrochloride salt used as a drug intermediate in medicinal chemistry research and chemical synthesis.

  • A chiral pyridinyl ethanamine used as a synthetic intermediate.
  • Appearance: light yellow to yellow solid.
  • Purity about 98.3% by assay.
  • CAS number 1980023-97-9, molecular formula C10H13Cl2FN4, molecular weight 279.15.
  • Available in small lab sizes from 100 mg to 10 g.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-10633 Supplier Medchemexpress LLC Supplier No. HYW068075A1G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A chiral dihydrochloride salt used as a drug intermediate in medicinal chemistry research and chemical synthesis.

  • A chiral pyridinyl ethanamine used as a synthetic intermediate.
  • Appearance: light yellow to yellow solid.
  • Purity about 98.3% by assay.
  • CAS number 1980023-97-9, molecular formula C10H13Cl2FN4, molecular weight 279.15.
  • Available in small lab sizes from 100 mg to 10 g.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.