Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-1-Amino-2-propanol, 98%, Thermo Scientific™

Catalog No. AAL14102MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL14102MD 250 mg
AAL1410203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL14102MD Supplier Thermo Scientific Chemicals Supplier No. L14102MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2799-17-9
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00064429
InChI Key HXKKHQJGJAFBHI-UHFFFAOYNA-N
Synonym s-+-1-amino-2-propanol, s-1-aminopropan-2-ol, 2s-1-aminopropan-2-ol, +-isopropanolamine, s-1-amino-2-propanol, 2-propanol, 1-amino-, 2s, 1-amino-2-propanol #, 2-propanol, 1-amino-, s, ambotzfal1033
PubChem CID 7311736
IUPAC Name 1-aminopropan-2-ol
SMILES CC(O)CN

Specifications

Melting Point 24°C to 26°C
Density 0.96
Boiling Point 160°C
Flash Point 71°C (159°F)
Refractive Index 1.449
Quantity 250 mg
UN Number UN3259
Beilstein 1718868
Sensitivity Air sensitive; Hygroscopic
Optical Rotation +18.5° (c=1.8 in Water)
Formula Weight 75.11
Percent Purity 98%
Chemical Name or Material (S)-(+)-1-Amino-2-propanol
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 220-533-4
RTECSNumber UA5775000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.