Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-1-Boc-3-aminopiperidine | 625471-18-3 | 99.8% | 200.28 | 25 G
SDP

Catalog No. 5000403095
Encompass_Preferred

(S)-1-Boc-3-aminopiperidine is a crucial intermediate utilized in the synthesis of various novel inhibitors, including CHK1 inhibitors and PI3Kδ inhibitors. It is intended for research use only and not for sale to patients.

  • Key intermediate for synthesizing novel inhibitors.
  • Used in the creation of CHK1 inhibitors.
  • Utilized in the creation of PI3Kδ inhibitors.
  • Available in various quantities.
  • Appearance: Solid-Liquid Mixture.
  • Color: Colorless to light yellow.
  • Soluble in DMSO (100 mg/mL).
  • Storage at 4°C, protected from light.
  • Stock solution storage: -80°C for 6 months; -20°C for 1 month (protect from light).

Catalog No. 50-004-03095 Supplier Medchemexpress LLC Supplier No. 25GHY40029
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-1-Boc-3-aminopiperidine is a crucial intermediate utilized in the synthesis of various novel inhibitors, including CHK1 inhibitors and PI3Kδ inhibitors. It is intended for research use only and not for sale to patients.

  • Key intermediate for synthesizing novel inhibitors.
  • Used in the creation of CHK1 inhibitors.
  • Utilized in the creation of PI3Kδ inhibitors.
  • Available in various quantities.
  • Appearance: Solid-Liquid Mixture.
  • Color: Colorless to light yellow.
  • Soluble in DMSO (100 mg/mL).
  • Storage at 4°C, protected from light.
  • Stock solution storage: -80°C for 6 months; -20°C for 1 month (protect from light).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.