Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-1-Boc-3-methylpiperazine | 147081-29-6 | 100.0% | 200.28 | 1 ML
SDP

Catalog No. 5000403841
Encompass_Preferred

S-1-Boc-3-methylpiperazine is a versatile drug intermediate primarily utilized in the synthesis of sotorasib. This compound is presented as a white to off-white solid, requiring specific handling and storage conditions to maintain its integrity for research applications.

  • Molecular weight: 200.28
  • Formula: C10H20N2O2
  • CAS number: 147081-29-6
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light; in solvent: -80°C for 6 months, -20°C for 1 month (protect from light)

Catalog No. 50-004-03841 Supplier Medchemexpress LLC Supplier No. HY4004010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S-1-Boc-3-methylpiperazine is a versatile drug intermediate primarily utilized in the synthesis of sotorasib. This compound is presented as a white to off-white solid, requiring specific handling and storage conditions to maintain its integrity for research applications.

  • Molecular weight: 200.28
  • Formula: C10H20N2O2
  • CAS number: 147081-29-6
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light; in solvent: -80°C for 6 months, -20°C for 1 month (protect from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.