Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-1-Boc-3-methylpiperazine | 147081-29-6 | 100.0% | 200.28 | 5 G
SDP

Catalog No. 5000405026
Encompass_Preferred

(S)-1-Boc-3-methylpiperazine is a drug intermediate primarily used for the synthesis of Sotorasib. This product is a solid, typically white to off-white in color. It is intended for research use only and has not been fully validated for medical applications.

  • High purity of 99.97%
  • Molecular weight of 200.28
  • Solid, white to off-white appearance
  • Soluble in DMSO at 100 mg/mL
  • Recommended storage at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • Identified by CAS number 147081-29-6

Catalog No. 50-004-05026 Supplier Medchemexpress LLC Supplier No. HY400405G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-1-Boc-3-methylpiperazine is a drug intermediate primarily used for the synthesis of Sotorasib. This product is a solid, typically white to off-white in color. It is intended for research use only and has not been fully validated for medical applications.

  • High purity of 99.97%
  • Molecular weight of 200.28
  • Solid, white to off-white appearance
  • Soluble in DMSO at 100 mg/mL
  • Recommended storage at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • Identified by CAS number 147081-29-6

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.