Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-1-Boc-3-Methylpiperazine

Catalog No. 632075GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-207-5GF 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 63-207-5GF Supplier Sigma Aldrich Supplier No. 632075GF
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant involved in the synthesis of biologically activy molecules including:CCR5 antagonists with anti-HIV-1 activityOpioid receptor antagonistsHuman growth hormone secretagogue receptor antagonists for treatment of obesityFatty acid oxidation inhibitors

Specifications

CAS 213843-90-4
Melting Point 218°C to 221°C
Molecular Formula C33H34NO4P
Linear Formula C33H34NO4P
MDL Number MFCD04117685
Quantity 500 mg
Synonym tert-Butyl (S)-3-methyl-1-piperazinecarboxylate
Molecular Weight (g/mol) 539.6 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.