Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-1-(Diphenylphosphino)-3,3-Dimethylbutan-2-Amine

Catalog No. 748307500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-830-7500MG 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 74-830-7500MG Supplier Sigma Aldrich Supplier No. 748307500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-1-(Diphenylphosphino)-3,3-Dimethylbutan-2-Amine

Specifications

CAS 10198-89-7
Melting Point 104°C to 109°C
Molecular Formula C13H10N2O2
Refractive Index n20/D 1.590
Linear Formula C13H10N2O2
MDL Number MFCD01321157
Quantity 5 g
Synonym 1,3-Bis(2-pyridyl)-1,3-propanedione; 1,3-Di-2-pyridinyl-1,3-propanedione
Molecular Weight (g/mol) 226.23 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.