Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-1-(Diphenylphosphino)-3-Methyl-2-Butylamine

Catalog No. 748277500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-827-7500MG 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 74-827-7500MG Supplier Sigma Aldrich Supplier No. 748277500MG
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-1-(Diphenylphosphino)-3-Methyl-2-Butylamine

Specifications

Density 1.334 g/mL at 25°C
Molecular Formula C3H9ISi
Refractive Index n20/D 1.597
Linear Formula C3H9ISi
Quantity 100 mL
Synonym (S)-1-(Diphenylphosphino)-3-methylbutan-2-amine
Molecular Weight (g/mol) 200.09 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.