Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(+)-1-Methoxy-2-Propanol

Catalog No. 888081MLF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-808-1MLF 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 88-808-1MLF Supplier Sigma Aldrich Supplier No. 888081MLF
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(+)-1-Methoxy-2-propanol can be used as a reactant to prepare: (S)-1-methoxypropan-2-yl 4-methylbenzenesulfonate by reacting with tosyl chloride in the presence of pyridine. Thiazolopyridine urea derivatives. Quinazoline derivatives as potential inhibitors of EGFR/HER-2 tyrosine kinases.

Specifications

CAS 223121-01-5
Molecular Formula C32H40FeP2
Refractive Index n20/D 1.403
Linear Formula C32H40FeP2
MDL Number MFCD08561171
Quantity 500 mg
Synonym (S)-(+)-Propylene glycol 1-methyl ether
Molecular Weight (g/mol) 542.45 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.