Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-1-Methoxy-2-Propylamine

Catalog No. 7272965G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-729-65G 5 g
72-729-625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 72-729-65G Supplier Sigma Aldrich Supplier No. 7272965G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-1-Methoxy-2-propylamine can be used:As an intermediate in the synthesis of piperazinebenzylamine based human MC4 receptor antagonists.To prepare imidazopyrimidine derivatives as potent p38 MAP kinase inhibitors.In the synthesis of a marine natural product nhatrangin A.ChIpr∗

Specifications

CAS 99636-32-5
Molecular Formula C4H11NO
Linear Formula C4H11NO
MDL Number MFCD01075730
Quantity 5 g
Synonym (S)-2-Amino-1-methoxypropane
Molecular Weight (g/mol) 89.14 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.